Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simulated Gastric Fluid (without enzyme), J.T.Baker™

Click to view available options
Quantity:
12x230.8mL
6x961.5mL
Packaging:
Nm Poly Bottle
Poly Bottle

Chemical Identifiers

7647-01-0
ClH
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
CAS 7647-01-0
Molecular Formula ClH
Density Estimated 1 (20°C)
Solubility Information Completely Soluble
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name chlorane
Molecular Weight (g/mol) 36.458
PubChem CID 313
ChEBI CHEBI:17883
Physical Form Liquid
Odor Acrid odor, sharp like hydrochloric acid
Packaging Poly Bottle
Color Colorless
pH 0.4
Quantity 12x230.8mL
Chemical Name or Material Simulated Gastric Fluid, without enzyme
Show More Show Less