Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sin3A-associated protein Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | Sin3A-associated protein |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB023993
|
Novus Biologicals
NBP294735100UL |
100 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB023994
|
Novus Biologicals
NBP29473520UL |
20 μg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Sin3A-associated protein Polyclonal antibody specifically detects Sin3A-associated protein in Mouse samples. It is validated for Western BlotSpecifications
Sin3A-associated protein | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.3), 50% glycerol | |
79595 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
130 kDa Sin3-associated polypeptide, histone deacetylase complex subunit SAP130, mSin3A-associated protein 130, SAP130, SAP130 Sin3A-associated protein, 130kDa, Sin3-associated polypeptide p130 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 819-1048 of human SAP130 (NP_078821.2). | |
Primary | |
Store at -20°:C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title