Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIN3B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | SIN3B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
SIN3B Polyclonal specifically detects SIN3B in Mouse samples. It is validated for Western Blot.Specifications
SIN3B | |
Polyclonal | |
Purified | |
RUO | |
Histone deacetylase complex subunit Sin3b, KIAA0700paired amphipathic helix protein Sin3b, SIN3 homolog B, transcription regulator (yeast), SIN3 homolog B, transcriptional regulator, SIN3 homolog B, transcriptional regulator (yeast), Transcriptional corepressor Sin3b | |
SIN3B | |
IgG | |
Protein A purified |
Western Blot | |
Unconjugated | |
Rabbit | |
NP_033214 | |
23309 | |
Synthetic peptide directed towards the N terminal of human SIN3B. Peptide sequence NIQSPLSSQDNSHSHGDCGEDFKQMSYKEDRGQVPLESDSVEFNNAISYV. | |
Primary | |
105 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title