Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sinapic acid Matrix Substance for MALDI-MS (T), ≥99.0% (T), MilliporeSigma™ Supelco™
Sinapic acid is a type of phenolic acid. Removal of amino group from phenylalanine, catalysing with enzyme phenylalanine ammonia-lyase forms cinnamic acid. The precursor of hydroxycinnamates is produced after hydroxylation of benzene ring. One of the precursors produced is sinapic acid.
$119.30 - $211.00
Specifications
CAS | 530-59-6 |
---|---|
Type | Matrix Substance for MALDI-MS |
Linear Formula | C11H12O5 |
MDL Number | MFCD00004401 |
Synonym | 3,5-Dimethoxy-4-hydroxycinnamic acid; 4-Hydroxy-3,5-dimethoxy-cinnamic acid; Sinapinic acid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-4479
|
MilliporeSigma Supelco
854291G |
1 g |
Each for $119.30
|
|
|||||
11-101-4480
|
MilliporeSigma Supelco
854295G |
5 g |
Each for $211.00
|
|
|||||
Specifications
530-59-6 | |
C11H12O5 | |
3,5-Dimethoxy-4-hydroxycinnamic acid; 4-Hydroxy-3,5-dimethoxy-cinnamic acid; Sinapinic acid | |
≥99.0% (T) | |
202°C |
Matrix Substance for MALDI-MS | |
MFCD00004401 | |
224.21 | |
Matrix Substance for MALDI-MS | |
Soluble in dioxane |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title