Learn More
Eisco Single Channel Variable Volume Micropipettes
Eisco Single Channel Variable Volume Micropipettes are designed to deliver precise liquid volumes.
Manufacturer: Eisco CH0582F
Description
- Variable volume micropipette
- Single channel
- Simple digital display volume adjustment
- 2 stop plunger for sample intake/dispense
- Mechanical tip ejector
- Air displacement technology
Specifications
10 μl | |
30 x 8 x 4 cm | |
Micropipette Tips | |
Micropipette | |
Red | |
1000 μL |
S39856, S39865, S39872 | |
Electrophoresis, PCR | |
Non-Sterile | |
1 Year | |
Plastic |