Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Single Sheet Metal Burette Clamp Products for Science Education

This single metal burette clamp is made from die-pressed sheet metal and includes a built in boss head. It can accommodate 1 burette (20 mm max. dia.) and can be mounted on a rod up to 10 mm in diameter with the screw adjustment. 4 vinyl covered fingers provide a strong and steady grip.

Manufacturer:  Eisco™ CH0254A

Catalog No. S40579

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

  • Made from die-pressed sheet metal
  • Includes built in boss head with screw adjusment
  • 4 vinyl covered fingers
  • Can hold 1 burette, 20 mm max. diameter
  • Can be mounted on a rod with 10 mm max. diameter

Warranty and Services

1 year limited manufacturer warranty

Specifications

Specifications

10 mm
1
Single Burette Clamp
Silver
Burette
Single Burette Clamp
1
(4) Fingers, Screw Adjustment
Steel
Titration
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit