Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AIMS™ Single Temperature UIHD-Power Pack for Warming Chamber

For use with any single temperature warming chamber.

Manufacturer:  AIMS™ PP34

 View more versions of this product

Catalog No. 14-370-310


Add to cart

Description

Description

  • Universal AC Input (110 to 220V) for use in any country
  • Power Pack has C14 socket for use with a C13 power cord end
  • Molded design for maximum durability and long life
Specifications

Specifications

Black
AIMS 34°C Single Temp Post-op Warming Chamber
4 x 2 x 1.5 in.
CE, UL, CCC, PCT
Molded ABS
rectangle
Universal Input 110-220V AC
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit