Learn More
Globe Scientific Single Thumb Tab Snap Caps
Colorful snap-on closures are an effective way to recap blood drawing tubes, glass test tubes and plastic culture tubes.
Manufacturer:  Globe Scientific 113139C
 View more versions of this product
Catalog No. 22-171-707
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Material: Low-density polyethylene (LDPE)
  • Features one thumb tab for easy removal of the cap
  • Allows more tubes to be stacked closely in racks
  • Maintain tight seal during centrifugation, agitation and refrigeration procedures
  • Available in bright attractive colors for sample identification purposes
Specifications
Specifications
Polyethylene
10 mm Blood drawing tubes, glass test tubes and plastic culture tubes
1000
Clear
Snap cap