Learn More
Globe Scientific Single Thumb Tab Snap Caps
Colorful snap-on closures are an effective way to recap blood drawing tubes, glass test tubes and plastic culture tubes.
Products 5
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
22-010-003
View Documents
Globe Scientific
113139B
Blue This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
22-010-004
View Documents
Globe Scientific
113139L
Lavender This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
22-010-005
View Documents
Globe Scientific
113139R
Red This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
22-010-006
View Documents
Globe Scientific
113139Y
Yellow This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
22-171-707
View Documents
Globe Scientific
113139C
Clear This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Material: Low-density polyethylene (LDPE)
  • Features one thumb tab for easy removal of the cap
  • Allows more tubes to be stacked closely in racks
  • Maintain tight seal during centrifugation, agitation and refrigeration procedures
  • Available in bright attractive colors for sample identification purposes