Learn More
Honeywell™ Single-Use Powder-Free Nitrile Gloves, 4 mil
High-quality nitrile gloves that are certified for food handling.
Manufacturer:  Honeywell™ 4580386XXL
 View more versions of this product
Catalog No. 19-085-582
Add to cart
Description
Description
  • AQL 1.5 and food handling certified
  • Guarantees a high level of quality and contact with foodstuffs (non-damaging to health, glove components do not migrate toward food)
  • Nitrile glove apply to those allergic to latex
  • Very fine touch
Specifications
Specifications
4 mil
2X-Large
9.5 in.
Nitrile
Blue