Sino Biological™ ACLY Recombinant Human Protein, N-His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the human ACLY (P53396) (Met 1-Met 1101) fused to a polyhistidine tag at the N-terminus

Manufacturer: Invitrogen 11769H07B5

 View more versions of this product

Catalog No. 11-769-H07B5

  N/A
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Gene ID (Entrez) 47
    Purification Method PAGE
    Purity ≥85% by SDS-PAGE
    Source Human
    Storage Requirements Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Endotoxin Concentration <1 EU/μg
    Gene Alias ACL, ATPCL, CLATP
    Gene Symbol ACLY
    Protein Tag His Tag
    Validated Application Assay Standard
    Expression System Insect
    Format Frozen
    Molecular Weight 123kDa
    Quantity 5μg
    Structural Form Full Length/Recombinant
    Common Name ATP Citrate Lyase
    Protein Family Other Proteins
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

    ATP Citrate Lyase (ACLY), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: This recombinant protein has not been tested
    • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, pH 8.0
    • Reconstitution: No reconstitution needed

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit