Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ APP Recombinant Human Protein, N-His.GST Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the amino acids (Asp 672-Val 711) of human Amyloid beta A4 protein (APP770) (P05067-1), corresponding to the Beta-amyloid protein 40, fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

Manufacturer:  Invitrogen 10703H20E125

 View more versions of this product

Catalog No. 10-703-H20E12


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Amyloid beta (A4) precursor (APP), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind biotinylated recombinant human AGER (Mfr. No. 11629-HCCH) in a functional ELISA
  • Formulation: Lyophilized in 50mM Tris, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
351
PAGE
5 x 5μg
N-Terminal
<1 EU/μg
Amyloid beta (A4) precursor protein
Assay Standard
E. coli
Lyophilized
31.8kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
Beta-APP40
APP
Other Proteins
GST Tag, His Tag
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit