Sino Biological™ APP Recombinant Human Protein, N-His.GST Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the amino acids (Asp 672-Val 711) of human Amyloid beta A4 protein (APP770) (P05067-1), corresponding to the Beta-amyloid protein 40, fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

Manufacturer: Invitrogen 10703H20E15

 View more versions of this product

Catalog No. 10-703-H20E15

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 351
    Purification Method PAGE
    Purity ≥90% by SDS-PAGE
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
    Termination N-Terminal
    Endotoxin Concentration <1 EU/μg
    Protein Tag GST Tag/His Tag
    Molecular Weight 31.8kDa
    Quantity 5μg
    Structural Form Full Length/Recombinant
    Gene Alias Beta-APP40
    Common Name Amyloid beta (A4) precursor protein
    Gene Symbol APP
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System E. coli
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Amyloid beta (A4) precursor (APP), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its ability to bind biotinylated recombinant human AGER (Mfr. No. 11629-HCCH) in a functional ELISA
    • Formulation: Lyophilized in 50mM Tris, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit