Sino Biological™ BMPR1A, Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc.His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 152) of human ALK3 (NP_004320.2) fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer: Invitrogen 10446H03H50

 View more versions of this product

Catalog No. 10-446-H03H50

  N/A
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Gene ID (Entrez) 657
    Molecular Weight 42kDa
    Purification Method PAGE
    Purity ≥85% by SDS-PAGE
    Quantity 50μg
    Structural Form Truncated/Recombinant
    Endotoxin Concentration <1 EU/μg
    Gene Symbol BMPR1A
    Protein Tag hIgG1-Fc His
    Protein Family Growth Factors & Receptors
    Expression System Mammalian
    Storage Requirements Store at -5° to -30°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Gene Alias ACVRLK3, ALK3, CD292, BMPR1A
    Common Name Bone morphogenetic protein receptor, type IA
    Validated Application Assay Standard
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

    Bone Morphogenetic Protein Receptor, Type IA (BMPR1A), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

    • Activity: Measured by its ability to inhibit BMP4 induced activity in MC3T3-E1 Mouse osteoblastic cells, the ED50 for this effect is typically 0.1-0.3µg/mL in the presence of 50ng/mL of recombinant human BMP4
    • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit