Sino Biological™ CADM3 Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the human CADM3 (short isoform) (NP_001120645.1) extracellular domain (Met 1-His 330) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 11214H08H25

 View more versions of this product

Catalog No. 11-214-H08H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Gene Symbol CADM3
    Protein Tag His Tag
    Validated Application Assay Standard
    Molecular Weight 35.1kDa
    Protein Subtype Neuropeptides|Intercellular Adhesion Molecule
    Endotoxin Concentration <1 EU/μg
    Gene Alias CADM3, CTA-134P22.1, BIgR, FLJ10698, IGSF4B, NECL1, Necl-1, TSLL1, synCAM3
    Common Name Cell adhesion molecule 3
    Protein Family Other Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Cell Adhesion Molecule 3 (CADM3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

    • Activity: Measured by the ability of the immobilized protein to support the adhesion of C6 Rat brain glial cells; human CADM3 immobilized at 0.8µg/mL, 100µL/well will mediate >30% C6 cell adhesion
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit