Sino Biological™ CCL1 Recombinant Mouse Protein, N-hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the secreted form of mouse CCL1 isoform 1 (P10146-1) (Lys 24-Cys 92) fused to the Fc region of human IgG1 at the N-terminus

Manufacturer: Invitrogen 50178M01H25

 View more versions of this product

Catalog No. 50-178-M01H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of protein in suspension buffer

    Specifications

    Gene ID (Entrez) 20290
    Molecular Weight 36.2kDa
    Purification Method PAGE
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
    Structural Form Full Length/Recombinant
    Protein Subtype Other Cytokines
    Gene Alias RP23-446K18.2, BF534335, I-309, Scya1, Tca-3
    Common Name Chemokine (C-C motif) ligand 1
    Gene Symbol CCL1
    Protein Tag IgG1 Fc Tag
    Validated Application Assay Standard
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized
    Termination N-Terminal
    Endotoxin Concentration <1 EU/μg
    Protein Family Chemokines & Receptors

    Chemokine (C-C Motif) Ligand 1 (CCL1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: The activity of this protein has not been tested
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit