Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD19 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ Non-distribution product as customer accommodation.

This recombinant protein is expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 291) of human CD19 (AAB60697) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Invitrogen 11880H08H50

 View more versions of this product

Catalog No. 11-880-H08H50


Add to cart

Description

Description

Cluster of Differentiation 19 (CD19) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in analysis of protein structure the study of protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.Background: Cluster of Differentiation 19 (CD19), is a single-pass type I membrane protein which contains two Ig-like C2-type (immunoglobulinlike) domains. CD19 forms a complex with CD21, CD81 and CD225 in the membrane of mature B cells. It interacts with VAV. It interacts with GRB2 and SOS when phosphorylated on Tyr-348 and/or Tyr-378. CD19 also interacts with PLCG2 when phosphorylated on Tyr-409. CD19 is an important pan B cell marker and co-stimulatory protein in humans and mice. CD19 is a hallmark differentiation antigen of the B cell lineage and positively regulates antigen receptor signal transduction in mature B cells. CD19 defines signaling thresholds for the Ag receptor as well as other cell surface receptors that regulate B lymphocyte selection, activation, and differentiation. Defects in CD19 are a cause of hypogammaglobulinemia.
  • N-terminal Sequence Analysis: Pro 20
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD19 at 2µg/mL (100µL/well) has been shown to bind biotinylated human CD9 with a linear range of 0.032-0.8µg/mL
  • Formulation: lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, and 0.02% Tween™-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. Manufactured by Sino Biological (China).

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
930
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Proteins
B4, mgC12802
CD19
CD Proteins
His Tag
Lyophilized
31.6kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD19 molecule
Assay Standard
Mammalian
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.