Sino Biological™ CD200R1 Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 238) of mouse CD200R1 (NP_067300.1) fused to a C-terminal polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 50209M08H25

 View more versions of this product

Catalog No. 50-209-M08H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 57781
    Molecular Weight 24.6kDa
    Purity ≥85% by SDS-PAGE
    Quantity 5 x 5μg
    Structural Form Truncated/Recombinant/Cell Surface Protein
    Endotoxin Concentration <1 EU/μg
    Gene Alias CD200R, Mox2r, OX2R
    Common Name CD200 receptor 1
    Protein Tag His Tag
    Protein Family CD Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Protein Subtype Glycoproteins
    Gene Symbol CD200R1
    Validated Application Assay Standard
    Format Lyophilized

    Cluster of Differentiation 200 receptor 1 (CD200R1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

    • Activity: Measured by its ability to bind mouse CD200 in a functional ELISA; immobilized mouse CD200-His at 0.2µg/well can bind mouse CD200R1-His with a linear range of 0.8-4.0µg/mL
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit