Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ CD28 Recombinant Mouse Protein, hIgG1-Fc.His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 149) of mouse CD28 precursor (NP_031668.3) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Manufacturer:  Invitrogen 50103M03H50

 View more versions of this product

Catalog No. 50-103-M03H50


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Cluster of Differentiation 28 (CD28), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by its binding ability in a functional ELISA; 1. Immobilized mouse CD86 at 20µg/mL (100µL/well) can bind mouse CD28 -Fc with a linear range of 6.4-800ng/mL; 2. Immobilized rat CD86 at 2µg/mL (100µL/well) can bind mouse CD28-Fc with a linear range of 6.4-160ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12487
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Glycoproteins
CD28, CH29-189K17.3
CD28
CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
43kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
CD28 antigen
Assay Standard
Mammalian
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit