Sino Biological™ CD4 Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Thr 394) of mouse CD4 (NP_038516.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 50134M08H5

 View more versions of this product

Catalog No. 50-134-M08H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 12504
    Structural Form Truncated/Recombinant/Cell Surface Protein
    Protein Subtype Glycoproteins
    Protein Tag His Tag
    Molecular Weight 42.8kDa
    Purity ≥95% by SDS-PAGE
    Quantity 5μg
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Endotoxin Concentration <1 EU/μg
    Gene Alias L3T4, Ly-4, CD4
    Common Name CD4 antigen
    Gene Symbol CD4
    Validated Application Assay Standard
    Protein Family CD Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Cluster of Differentiation 4 (CD4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

    • Activity: Measured by the ability of the immobilized protein to support the adhesion of HeLa human cervical epithelial carcinoma cells; when 5 x 104 cells/well are added to CD4 coated plates (2.5µg/mL and 100µL/well), approximately 40-60 % will adhere specifically after 60 minutes at 37°C
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit