Sino Biological™ CD86 (B7-2) Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-His 239) of human CD86 (NP_008820.2) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 10699H08H250

 View more versions of this product

Catalog No. 10-699-H08HL

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 942
    Molecular Weight 26.2kDa
    Quantity 5 x 50μg
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Endotoxin Concentration <1 EU/μg
    Gene Alias B7-2, B70, CD28LG2, LAB72, mgC34413
    Common Name CD86 molecule
    Gene Symbol CD86
    Protein Tag His Tag
    Validated Application Assay Standard
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized
    Purity ≥90% by SDS-PAGE
    Structural Form Truncated/Recombinant/Cell Surface Protein
    Protein Subtype Other Proteins
    Protein Family CD Proteins

    Cluster of Differentiation 86 (CD86/B7-2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, studying protein-protein interactions, and cell biology research applications. It can also be used as an immunogen, or as a protein standard.

    • Activity: Measured by its binding ability in a functional ELISA; immobilized human CD86 at 20µg/mL (100µL/well) can bind human CD28 with a linear range of 32-800ng/mL
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit