Sino Biological™ CDH17 Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the human CDH17 (Q12864-1) extracellular domain (Met 1-Met 787) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 11360H08H50

 View more versions of this product

Catalog No. 11-360-H08H50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Quantity 50μg
    Gene Symbol CDH17
    Molecular Weight 87kDa
    Purity ≥85% by SDS-PAGE
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Protein Subtype Intercellular Adhesion Molecule
    Endotoxin Concentration <1 EU/μg
    Gene Alias CDH16, FLJ26931, HPT-1, HPT1, mgC138218, mgC142024
    Common Name Cadherin 17, LI cadherin (liver-intestine)
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Cell Structure/Organelle Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Cadherin 17, LI cadherin (liver-intestine) (CDH17), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

    • Activity: Measured by the ability of the immobilized protein to support the adhesion of MCF-7 human breast adenocarcinoma cells; when 5 x 104 cells/well are added to recombinant human Cadherin 17 coated plates (0.8µg/mL with 100µL/well), approximately >50% will adhere after 1 hour at 37°C
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit