Sino Biological™ CES2 Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CES2C (NP_663578.1) (Met 1-Lys 557) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 50515M08H250

 View more versions of this product

Catalog No. 50-515-M08HL

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

    Specifications

    Molecular Weight 60.4kDa
    Purity >85% by SDS-PAGE
    Quantity 5 x 50μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Protein Subtype Other Proteins
    Endotoxin Concentration <1 EU/μg
    Gene Alias ces2A3
    Common Name Carboxylesterase 2
    Gene Symbol CES2
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Carboxylesterase 2C (CES2C), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its ability to hydrolyze p-nitrophenylacetate; the specific activity is >90nM/min./µg
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit