Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ CSF1R (MCSFR) Recombinant Mouse Protein (without Catalytic Activity Domain), His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 511) of mouse CSF1R (NP_001032948.2) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Invitrogen 50059M08H250

 View more versions of this product

Catalog No. 50-059-M08HL


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Colony stimulating factor 1 receptor (CSF1R/MCSFR), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to bind recombinant mouse IL34 in a functional ELISA; immobilized mouse CSF1R-His at 10µg/mL (100µL/well) can bind biotinylated human CSF1-His; the ED50 of biotinylated human CSF1-His is 0.50µg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
12978
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Cytokine Receptors
ICsf1r, AI323359, CD115, CSF-1R, Csfmr, Fim-2, Fms, M-CSFR
CSF1R
Growth Factors & Receptors
His Tag
Lyophilized
56.8kDa
5 x 50μg
Recombinant/Cell Surface Protein
<1 EU/μg
Colony stimulating factor 1 receptor
Assay Standard
Mammalian
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit