Sino Biological™ CSF3R (CD114) Recombinant Human Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 621) of human CSF3R precursor (NP_000751.1) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: Invitrogen 10218H02H250

 View more versions of this product

Catalog No. 10-218-H02HL

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 1441
    Molecular Weight 93.3kDa
    Purity ≥95% by SDS-PAGE
    Quantity 5 x 50μg
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Truncated/Recombinant/Cell Surface Protein
    Protein Subtype Cytokine Receptors
    Endotoxin Concentration <1 EU/μg
    Gene Alias CD114, GCSFR, CSF3R
    Common Name Colony stimulating factor 3 receptor (granulocyte)
    Gene Symbol CSF3R
    Protein Tag IgG1 Fc Tag
    Validated Application Assay Standard
    Protein Family Cytokines & Receptors/CD Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Colony stimulating factor 3 receptor (CSF3R/CD114), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard or in other research applications.

    • Activity: Measured by its ability to inhibit the GCSF-induced proliferation of NFS-60 mouse myeloid cells, the ED50 for this effect is typically 1-4ng/mL in the presence of 0.125ng/mL of recombinant human GCSF
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit