Sino Biological™ DDC Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the full length of human DDC (NP_000781.1) (Met 1-Glu 480) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 10560H08B25

 View more versions of this product

Catalog No. 10-560-H08B25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 55kDa
    Quantity 5 x 5μg
    Storage Requirements Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Endotoxin Concentration <1 EU/μg
    Gene Alias AADC, DDC
    Common Name Dopa Decarboxylase
    Gene Symbol DDC
    Protein Tag His Tag
    Validated Application Assay Standard
    Expression System Insect
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Frozen
    Purity ≥85% by SDS-PAGE
    Protein Subtype Other Proteins

    Dopa Decarboxylase (DDC), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its ability to convert the substrate 3,4-dihydroxy L-phenylalanine (L-Dopa) to 3,4-dihydroxyphenylethylamine (dopamine); the dopamine product is measured by its absorbance at 340nM after derivatization with trinitrobenzene sulfonic acid, the specific activity is >1500pmole/min./µg
    • Formulation: 50mM Tris, 100mM NaCl, 0.2µg/µL BSA, pH 8, 10% glycerol
    • Reconstitution: No reconstitution needed

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit