Sino Biological™ DPP4 Recombinant Human Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Asn 29-Pro 766) of the mature form of human DPP4 (NP_001926.2) fused to the fused Fc region of human IgG1 at the N-terminus

Manufacturer: Invitrogen 10688H01H5

 View more versions of this product

Catalog No. 10-688-H01H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Common Name DPP4–dipeptidyl-peptidase 4
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Molecular Weight 112kDa
    Purity ≥95% by SDS-PAGE
    Quantity 5μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Protein Subtype Proteases
    Endotoxin Concentration <1 EU/μg
    Gene Alias DPP4, ADABP, ADCP2, CD26, DPPIV, TP103
    Gene Symbol DPP4
    Protein Tag IgG1 Fc Tag
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System Mammalian
    Format Lyophilized

    Dipeptidyl-Peptidase 4 (DPP4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: 1. Measured by its ability to bind recombinant Cynomolgus CXCL12 in a functional ELISA; 2.Measured by its ability to bind recombinant Human SDF1b in a functional ELISA
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit