Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ EFNA4 (EphrinA4) Recombinant Mouse Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the (Met 1-Gly 176) of mouse EFNA4 (NP_031936.2) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer:  Invitrogen 50595M02H50

 View more versions of this product

Catalog No. 50-595-M02H50


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Ephrin A4 (EFNA4), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human EPHA7 at 2µg/mL (100µL/well) can bind mouse EFNA4-Fc with a linear range of 0.08-10ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
13639
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Ephrins
AW494418, EFL-2, Ehk1-L, Epl3, LERK-3, mgC129300, mgC129301
EFNA4
Kinases & Inhibitors
IgG1 Fc Tag
Lyophilized
43.9kDa
50μg
Recombinant/Cell Surface Protein/Ligand
<1 EU/μg
Ephrin A4
Assay Standard
Mammalian
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit