Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ EGF Recombinant Canine Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mature form (Asn 973-Arg 1024) of canine EGF (Q9BEA0) fused to a polyhistidine tag at the N-terminus

Manufacturer:  Invitrogen 70013D07E50

 View more versions of this product

Catalog No. 70-013-D07E50


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Epidermal growth factor (EGF), recombinant canine protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: Measured in a cell proliferation assay using BALB/c 3T3 mouse embryonic fibroblasts; the ED50 for this effect is typically 0.1-0.5ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
403657
≥95% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
EGF (Epidermal Growth Factor)
Epidermal growth factor
Assay Standard
E. coli
Lyophilized
8.3kDa
50μg
Recombinant/Ligand
CEGF
EGF
Growth Factors & Receptors
His Tag
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit