Sino Biological™ EGFR Recombinant Mouse Protein (without Catalytic Activity Domain), hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse EGFR (Q01279) (Met 1-Ser 647) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: Invitrogen 51091M02H5

 View more versions of this product

Catalog No. 51-091-M02H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: One vial of lyophilized protein

    Specifications

    Molecular Weight 96kDa
    Purity ≥85% by SDS-PAGE
    Quantity 5μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Truncated/Recombinant/Cell Surface Protein
    Endotoxin Concentration <1 EU/μg
    Gene Alias RP23-295E4.1, 9030024J15Rik, AI552599, Erbb, Errp, Wa5, wa-2, wa2
    Protein Tag IgG1 Fc Tag
    Validated Application Assay Standard
    Protein Family Growth Factors & Receptors
    Expression System Mammalian
    Protein Subtype EGF (Epidermal Growth Factor)|Receptor Tyrosine Kinases
    Common Name Epidermal growth factor receptor
    Gene Symbol EGFR
    KinaseFamily EGF Receptor (ErbB) Kinase Family
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Epidermal Growth Factor Receptor (EGFR), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

    • Activity: Measured by its ability to bind human EGF in a functional ELISA
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween-80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit