Sino Biological™ EIF4EBP1 Recombinant Human Protein, N-His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the human 4EBP1 (NP_004086.1) (Ser 2-Ile 118) fused to a polyhistidine tag at the N-terminus

Manufacturer: Invitrogen 10022H07E50

 View more versions of this product

Catalog No. 10-022-H07E50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 1978
    Quantity 50μg
    Storage Requirements Store at -5° to -30°C. Store aliquots under sterile conditions at -5° to -30°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Full Length/Recombinant
    Validated Application Assay Standard
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Molecular Weight 13.4kDa
    Purification Method PAGE
    Purity ≥85% by SDS-PAGE
    Gene Alias 4E-BP1, EIF4EBP1, BP-1, mgC4316, PHAS-I
    Common Name Eukaryotic Translation Initiation Factor 4E Binding Protein 1
    Gene Symbol EIF4EBP1
    Protein Tag N-His
    Protein Family Other Proteins
    Expression System E. coli
    Format Frozen

    Eukaryotic Translation Initiation Factor 4E Binding Protein 1 (EIF4EBP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: This recombinant protein has not been tested
    • Formulation: 20mM Tris, 500mM NaCl, 10% glycerol, pH 8.0
    • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit