Sino Biological™ EPOR Recombinant Human Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met1-Pro 250) of human EPOR (NP_000112.1) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: Invitrogen 10707H02H250

 View more versions of this product

Catalog No. 10-707-H02HL

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 2057
    Molecular Weight 51.0kDa
    Quantity 5 x 50μg
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Truncated/Recombinant
    Protein Subtype Cytokine Receptors
    Endotoxin Concentration <1 EU/μg
    Gene Alias EPOR, mgC138358
    Gene Symbol EPOR
    Protein Tag IgG1 Fc Tag
    Validated Application Assay Standard
    Protein Family Hormones & Receptors
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized
    Purity ≥85% by SDS-PAGE
    Common Name Erythropoietin Receptor

    Erythropoietin receptor (EPOR), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its binding ability in a functional ELISA; immobilized CSF2RB/CD131 at 10µg/mL (100µL/well) can bind biotinylated recombinant human EPOR with a linear range of 0.16-4µg/mL
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit