Sino Biological™ FCGR1 (CD64) Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 297) of mouse FCGR1 precursor (NP_034316.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 50086M08H50

 View more versions of this product

Catalog No. 50-086-M08H50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 14129
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Molecular Weight 32kDa
    Purity ≥90% by SDS-PAGE
    Quantity 50μg
    Structural Form Truncated/Recombinant/Cell Surface Protein
    Protein Subtype Glycoproteins|Fc Receptors
    Endotoxin Concentration <1 EU/μg
    Gene Alias FCGR1, AI323638, AV092959, CD64, FcgammaRI, IGGHAFC
    Common Name Fc receptor, IgG, high affinity I
    Gene Symbol FCGR1
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Growth Factors & Receptors/CD Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Fc gamma receptor I (FCGR1/CD64), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: This recombinant protein has not been tested
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit