Sino Biological™ FETUB Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the human FETUB (NP_055190.2) (Met 1-Pro 382) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 11834H08H25

 View more versions of this product

Catalog No. 11-834-H08H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 26998
    Molecular Weight 41.7kDa
    Quantity 5 x 5μg
    Termination C-Terminal
    Structural Form Full Length/Recombinant
    Protein Subtype Kinase Inhibitors|Cytoplasmic Tyrosine Kinases|Protease Inhibitors
    Endotoxin Concentration <1 EU/μg
    Gene Alias 16G2, Gugu, IRL685
    Common Name Fetuin B
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Kinases & Inhibitors/Proteases, Peptidases & Inhibitors
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized
    Purification Method PAGE
    Purity ≥90% by SDS-PAGE
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
    Gene Symbol FETUB
    Expression System Mammalian

    Fetuin B (FETUB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: The activity of this protein has not been tested
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit