Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ FGF1 (aFGF) Recombinant Mouse Protein Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mature form (Phe 16-Asp 155) of mouse FGF1 (NP_034327.1)

Manufacturer:  Invitrogen 50179MNAE50

 View more versions of this product

Catalog No. 50-179-MNAE50


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Fibroblast growth factor 1 (FGF1/aFGF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard or in other research applications.
  • Activity: Measured in a cell proliferation assay using BALB/c 3T3 mouse fibroblasts; the ED50 for this effect is typically 0.05-0.25ng/mL in the presence of 10µg/mL heparin
  • Formulation: Lyophilized in 20mM MES buffer, pH 6.0, 200mM NaCl, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
14164
>98% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
FGFs (Fibroblast Growth Factors)
Fibroblast growth factor 1
Assay Standard
E. coli
Lyophilized
16kDa
50μg
Recombinant
Dffrx, Fam, Fgf-1, Fgfa, FGF1
FGF1
Growth Factors & Receptors
Untagged
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit