Sino Biological™ FGFR1 Recombinant Human Protein (without Catalytic Activity Domain), His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 285) of human FGFR1 (NP_075594.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 10616H08H250

 View more versions of this product

Catalog No. 10-616-H08HL

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 2260
    Molecular Weight 31kDa
    Purity >98% by SDS-PAGE
    Quantity 5 x 50μg
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Truncated/Recombinant
    Protein Subtype Receptor Tyrosine Kinases|Growth Factor Receptors
    Endotoxin Concentration <1 EU/μg
    Gene Alias FGFR1, BFGFR, CD331, CEK, FGFBR, FLG, FLJ99988, FLT2, HBGFR, KAL2, N-SAM
    Common Name Fibroblast growth factor receptor 1
    Gene Symbol FGFR1
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Growth Factors & Receptors
    Expression System Mammalian
    KinaseFamily FGF Receptor Kinase Family
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Fibroblast growth factor receptor 1 (FGFR1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

    • Activity: Measured by its ability to inhibit FGF1-dependent proliferation of BALB/c 3T3 mouse fibroblasts, the ED50 for this effect is typically 8-40ng/mL
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit