Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ FGFR1 Recombinant Mouse Protein (without Catalytic Activity Domain), hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 376) of mouse FGFR1 isoform 1 (P16092-1) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer:  Invitrogen 50186M02H250

 View more versions of this product

Catalog No. 50-186-M02HL


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Fibroblast growth factor receptor 1 (FGFR1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its ability to inhibit FGF1-dependent proliferation of BABL/c 3T3 mouse embryonic fibroblasts; the ED50 for this effect is typically 0.1-0.6ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
14182
≥85% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Receptor Tyrosine Kinases|Growth Factor Receptors
AW208770, FLG, Fgfr-1, Flt-2
FGFR1
Growth Factors & Receptors
FGF Receptor Kinase Family
Lyophilized
66.5kDa
5 x 50μg
Truncated/Recombinant
<1 EU/μg
Fibroblast growth factor receptor 1
Assay Standard
Mammalian
IgG1 Fc Tag
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit