Sino Biological™ FGFR1 Recombinant Mouse Protein (without Catalytic Activity Domain), His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 376) of mouse FGFR1 isoform 1 (P16092-1) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 50186M08H250

 View more versions of this product

Catalog No. 50-186-M08HL

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 14182
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Gene Alias AW208770, FLG, Fgfr-1, Flt-2
    Common Name Fibroblast growth factor receptor 1
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Molecular Weight 40.9kDa
    Purity ≥85% by SDS-PAGE
    Quantity 5 x 50μg
    Structural Form Truncated/Recombinant
    Protein Subtype Receptor Tyrosine Kinases|Growth Factor Receptors
    Endotoxin Concentration <1 EU/μg
    Gene Symbol FGFR1
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Growth Factors & Receptors
    Expression System Mammalian
    KinaseFamily FGF Receptor Kinase Family
    Format Lyophilized

    Fibroblast growth factor receptor 1 (FGFR1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

    • Activity: Measured by its ability to inhibit FGF1-dependent proliferation of BABL/c 3T3 mouse embryonic fibroblasts; the ED50 for this effect is typically 50-200ng/mL
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit