Sino Biological™ FGFR4 Recombinant Human Protein (without Catalytic Activity Domain), hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asp 369) of human FGFR4 precursor (NP_002002.3) fused to the Fc region of human IgG1 at the C-terminus

Manufacturer: Invitrogen 10538H02H5

 View more versions of this product

Catalog No. 10-538-H02H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 2264
    Protein Subtype FGFs (Fibroblast Growth Factors)|Receptor Tyrosine Kinases
    Endotoxin Concentration <1 EU/μg
    Gene Alias FGFR4, CD334, JTK2, mgC20292, TKF
    Common Name Fibroblast growth factor receptor 4
    Gene Symbol FGFR4
    Protein Tag IgG1 Fc Tag
    Validated Application Assay Standard
    Expression System Mammalian
    KinaseFamily FGF Receptor Kinase Family
    Molecular Weight 66kDa
    Purity ≥85% by SDS-PAGE
    Quantity 5μg
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Truncated/Recombinant
    Protein Family Growth Factors & Receptors
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    Fibroblast growth factor receptor 4 (FGFR4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

    • Activity: Measured by its ability to inhibit FGF1-dependent proliferation of BALB/c 3T3 mouse fibroblasts, the ED50 for this effect is typically 2-6ng/mL
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit