Sino Biological™ FH Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mature form of human FH (P07954-1) (Ala 45-Lys 510) fused to a polyhistidine tag at the C-terminus and an initial Met at the N-terminus

Manufacturer: Invitrogen 12115H08E50

 View more versions of this product

Catalog No. 12-115-H08E50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 2271
    Purification Method PAGE
    Purity >85% by SDS-PAGE
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Full Length/Recombinant
    Common Name Fumarate Hydratase
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System E. coli
    Format Lyophilized
    Molecular Weight 52kDa
    Quantity 50μg
    Termination C-Terminal
    Gene Alias HLRCC, LRCC, MCL, MCUL1
    Gene Symbol FH
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

    Fumarate Hydratase (FH), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

    • Activity: This recombinant protein has not been tested
    • Formulation: 10mM Tris, 5mM EDTA, 1mM DTT, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit