Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ FIGF Recombinant Mouse Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse FIGF (P97946) (Phe 98-Ser 206) fused to the Fc region of human IgG1 at the N-terminus

Manufacturer:  Invitrogen 50157M01H25

 View more versions of this product

Catalog No. 50-157-M01H25


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

Specifications

Description

C-fos induced growth factor (vascular endothelial growth factor D) (FIGF/VEGFD), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human VEGFR3-His at 10µg/mL (100µL/well) can bind mouse Fc-VEGFD; the EC50 of mouse VEGFD-Fc is 49ng/mL; 2. Immobilized mouse VEGFR3-His at 10µg/mL (100µL/well) can bind mouse Fc-VEGFD, the EC50 of mouse VEGFD-Fc is 44ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
40.6kDa
5 x 5μg
Full Length/Recombinant
<1 EU/μg
c-fos induced growth factor
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Growth Factors
RP23-202H24.2, AI325264, VEGF-D, Vegfd
FIGF
Growth Factors & Receptors
IgG1 Fc Tag
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit