Sino Biological™ FIGF Recombinant Mouse Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse FIGF (P97946) (Phe 98-Ser 206) fused to the Fc region of human IgG1 at the N-terminus

Manufacturer: Invitrogen 50157M01H250

 View more versions of this product

Catalog No. 50-157-M01HL

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

    Specifications

    Molecular Weight 40.6kDa
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 50μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Full Length/Recombinant
    Protein Subtype Other Growth Factors
    Endotoxin Concentration <1 EU/μg
    Gene Alias RP23-202H24.2, AI325264, VEGF-D, Vegfd
    Common Name c-fos induced growth factor
    Gene Symbol FIGF
    Protein Tag IgG1 Fc Tag
    Validated Application Assay Standard
    Protein Family Growth Factors & Receptors
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized
    Expression System Mammalian

    C-fos induced growth factor (vascular endothelial growth factor D) (FIGF/VEGFD), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human VEGFR3-His at 10µg/mL (100µL/well) can bind mouse Fc-VEGFD; the EC50 of mouse VEGFD-Fc is 49ng/mL; 2. Immobilized mouse VEGFR3-His at 10µg/mL (100µL/well) can bind mouse Fc-VEGFD, the EC50 of mouse VEGFD-Fc is 44ng/mL
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit