Sino Biological™ FKBP1A (FKBP12) Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the human FKBP12 (NP_463460) (Met 1-Glu 108) fused to a C-terminal polyhistidine tag

Manufacturer: Invitrogen 10268H08E250

 View more versions of this product

Catalog No. 10-268-H08EL

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Gene ID (Entrez) 2280
    Molecular Weight 12.9kDa
    Purification Method PAGE
    Purity ≥95% by SDS-PAGE
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Termination C-Terminal
    Gene Alias FKBP1A, RP11-314N13.2, FKBP-12, FKBP1, FKBP12, FKBP12C, PKC12, PKCI2, PPIASE
    Gene Symbol FKBP1A
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Proteases, Peptidases & Inhibitors/Phosphatase Inhibitors
    Format Lyophilized
    Quantity 5 x 50μg
    Structural Form Full Length/Recombinant
    Common Name FK506 binding protein 1A, 12kDa
    Expression System E. coli
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories

    FK506 binding protein 1A, 12kDa (FKBP1A), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

    • Activity: This recombinant protein has not been tested
    • Formulation: 20mM HEPES, 0.1 M NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit