Sino Biological™ FN1 Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the Fragment 2 (Ser 607 - Pro 1265) of human FN1 (CAD91166) fused to a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 10314H08H50

 View more versions of this product

Catalog No. 10-314-H08H50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 73.2kDa
    Purity ≥90% by SDS-PAGE
    Quantity 50μg
    Structural Form Truncated/Recombinant
    Protein Subtype Fibronectin
    Gene Alias FN, CIG, FNZ, MSF, ED-B, FINC, GFND, LETS, GFND2
    Gene Symbol FN1
    Protein Tag His Tag
    Protein Family Other Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Endotoxin Concentration <1 EU/μg
    Common Name Fibronectin 1
    Validated Application Assay Standard
    Format Lyophilized

    Fibronectin 1 (FN1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse APCS at 10µg/mL (100µL/well) can bind biotinylated FN1 with a linear range of 0.625 - 5µg/mL
    • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit