Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GFP Recombinant Aequorea victoria Protein, N-His Tag, Invitrogen™ Sino Biological™ Non-distribution product as customer accommodation.

This recombinant protein is expressed from a DNA sequence encoding the Aequorea victoria GFP (AAB65663) (Ser 2-Lys 238) fused to a polyhistidine tag at the N-terminus

Specifications

Purity ≥85% by SDS-PAGE
Purification Method PAGE
Format Lyophilized
Gene Alias GFP
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
13-105-S07E50 Non-distribution product as customer accommodation.
View Documents
Invitrogen
13105S07E50
50μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
13-105-S07EL Non-distribution product as customer accommodation.
View Documents
Invitrogen
13105S07E250
5 x 50μg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Green Fluorescent Protein (GFP) recombinant Aequorea victoria protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein structure analysis and in cell biology research applications.
Background: The green fluorescent protein (GFP) is a protein that exhibits bright green fluorescence when exposed to blue light. The protein is in the shape of a cylinder, comprising 11 strands of β-sheet with an α-helix inside and short helical segments on the ends of the cylinder. Its ability to generate a highly visible, efficiently emitting internal fluorophore is both intrinsically fascinating and tremendously valuable. The green-fluorescent protein (GFP) of the jellyfish Aequorea victoria has long been used as a universal reporter in a broad range of heterologous living cells and organisms. GFP has become well established as a marker of gene expression and protein targeting in intact cells and organisms.
  • N-terminal Sequence Analysis: Glu 99
  • Activity: 1. Mesured by emission analysis; 2. Measure excitation and emission spectra of GFP at wavelengths of 395nM and 510nM respectively
  • Formulation: Lyophilized from sterile PBS, pH 7.5
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Manufactured by Sino Biological (China).

Specifications

Specifications

≥85% by SDS-PAGE
PAGE
Lyophilized
GFP
SDS
Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit