Sino Biological™ HA Recombinant Influenza A, Subtype H1N1 (A/England/195/2009) Protein, His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of the influenza A H1N1 (A/England/195/2009 (H1N1)) hemagglutinin (ACR15621.1) (Met 1-Gln 529, HA1+HA2, uncleved) fused to a C-terminal polyhistidine tag

Manufacturer: Invitrogen 40005V08H25

 View more versions of this product

Catalog No. 40-005-V08H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Termination C-Terminal
    Endotoxin Concentration <1 EU/μg
    Gene Alias HA
    Protein Tag His Tag
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Molecular Weight 59kDa
    Purification Method PAGE
    Structural Form Truncated/Recombinant
    Common Name Hemagglutinin
    Gene Symbol HA
    Validated Application Assay Standard
    Protein Family Cell Structure/Organelle Proteins
    Format Lyophilized

    Hemagglutinin (HA), recombinant viral protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein stucture analysis and in cell biology research applications.

    • Activity: This recombinant protein has not been tested
    • Formulation: 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit