Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ HA Recombinant Influenza A Virus Protein Subtype H4N8 (A/Chicken/Alabama/1/1975), His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of influenza A virus hemagglutinin (A/chicken/Alabama/1/1975(H4N8) P19695 (Met 1-Ile528) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Invitrogen 40025V08H50

 View more versions of this product

Catalog No. 40-025-V08H50


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Hemagglutinin A (HA), recombinant H4N8 (A/chicken/Alabama/1/1975) HA protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Unknown
PAGE
50μg
C-Terminal
<1 EU/μg
Hemagglutinin A
Assay Standard
Mammalian
Lyophilized
59kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Truncated/Recombinant
HA
HA
Cell Structure/Organelle Proteins
His Tag
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit