Sino Biological™ HA1 Recombinant Influenza A Virus Protein Subtype H1N1 (A/England/195/2009), His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the N-terminal segment (Met 1-Arg 344) of influenza A H1N1 (A/England/195/2009 (H1N1)) hemagglutinin (ACR15621.1) termed as HA1, and this protein has a polyhistidine tag at the C-terminus

Manufacturer: Invitrogen 40005V08H125

 View more versions of this product

Catalog No. 40-005-V08H12

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Purification Method PAGE
    Purity ≥90% by SDS-PAGE
    Quantity 5 x 5μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Termination C-Terminal
    Structural Form Truncated/Recombinant
    Protein Subtype Viral Proteins|Glycoproteins
    Endotoxin Concentration <1 EU/μg
    Gene Alias HA1
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family Cell Structure/Organelle Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized
    Gene ID (Entrez) Unknown
    Molecular Weight 37.7kDa
    Common Name Hemagglutinin A, subunit 1
    Gene Symbol HA1

    Hemagglutinin A, Subunit 1 (HA1), recombinant Influenza A H1N1 (A/England/195/2009) HA1 protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

    • Activity: The activity of this protein has not been tested
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit