Sino Biological™ HA1 Subunit Recombinant Influenza A Virus Protein, Subtype H5N1, (A/Vietnam/1203/2004), mIgG1-Fc.His Tag Non-distribution product as customer accommodation.

This recombinant protein was expressed from a DNA sequence encoding the N-terminal segment (Met 1-Arg 342) of influenza A virus, subtype H5N1 (A/VietNam/1203/2004), HA1 subunit (AAW80717.1) fused to the His-tagged Fc region of mouse IgG1 at the C-terminus

Manufacturer: Invitrogen 10003V06H125

 View more versions of this product

Catalog No. 10-003-V06H12

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 26.1kDa
    Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Structural Form Truncated/Recombinant/Cell Surface Protein
    Endotoxin Concentration <1 EU/μg
    Gene Alias HA1
    Common Name Hemagglutinin
    Protein Tag His Tag/IgG1 Fc Tag
    Validated Application Assay Standard
    Protein Family Other Proteins
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Purity ≥85% by SDS-PAGE and SEC-HPLC Analysis
    Quantity 5 x 5μg
    Protein Subtype Viral Proteins
    Gene Symbol HA
    Format Lyophilized

    H5N1, recombinant influenza A virus protein is supplied as a lyophilized powder. It is suitable for use in studying protein-protein interactions, and cell biology research applications. It can also be used as a protein standard.

    • Activity: Measured by its binding ability in a functional ELISA
    • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
    • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

    Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit