Sino Biological™ HMGB1 Recombinant Human Protein, hIgG1-Fc Tag Non-distribution product as customer accommodation.

This recombinant protein is expressed from a DNA sequence encoding the human HMGB1 protein (NP_002119.1) (Met 1-Glu 215) fused to the fused Fc region of human IgG1 at the N-terminus

Manufacturer: Invitrogen 10326H01H50

 View more versions of this product

Catalog No. 10-326-H01H50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 51.5kDa
    Purity ≥90% by SDS-PAGE
    Quantity 50μg
    Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    Protein Subtype Other Cytokines
    Endotoxin Concentration <1 EU/μg
    Gene Alias DKFZp686A04236, HMG1, HMG3, SBP-1, HMGB1
    Common Name High mobility group box 1
    Gene Symbol HMGB1
    Protein Tag IgG1 Fc Tag
    Validated Application Assay Standard
    Protein Family Cytokines & Receptors
    Expression System Mammalian
    For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    Format Lyophilized

    High Mobility Group Box 1 (HMGB1) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.Background: Human High-Mobility Group Box 1 protein (HMGB1), also known as HMG-1 or amphoterin previously, is a member of the HMGB family consisting of three members, HMGB1, HMGB2 and HMGB3. HMGB1 is a nonhistone architectural chromosomal protein ubiquitously present in all vertebrate nuclei and binds double-stranded DNA without sequence specificity. HMGB1 is structurally composed of two homologous DNA-binding domains entitled box A and box B and a highly negative-charged C terminus. This protein is extremely conserved across species with 99- 100% aa sequence homology to mouse, rat, bovine, canine, and porcine proteins. As a nuclear protein, HMGB1 stabilizes nucleosomes and allows bending of DNA that facilitates gene transcription which is essential for individual survival. Meanwhile, it is revealed that HMGB1 can also act as a cytokine extracellularly and regulates monocyte, T cell, dendritic cell activities in inflammatory responses. Once secreted, HMGB1 induces signal transduction through its receptor RAGE and perhaps TLR2 and TLR4. The immune-regulatory function of HMGB1 may lead to being identified as new therapeutic target.

    • N-terminal Sequence Analysis: Glu 20
    • Activity: measured by its binding ability in a functional ELISA; immobilized recombinant mouse AGER at 2µg/mL (100µL/well) has been shown to bind human HMGB1, the EC50 of human HMGB1 is 0.23µg/mL
    • Formulation: Reconstitution: dissolve the protein in sterile double-distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

    Manufactured by Sino Biological (China).

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit